Skip to Content
Merck
All Photos(1)

Key Documents

A8082

Sigma-Aldrich

Antifoam O-30

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161901

form

emulsion (aqueous)

Quality Level

concentration

100% (organic)

technique(s)

cell culture | hybridoma: suitable
microbiological culture: suitable

density

0.98 g/mL at 25 °C

storage temp.

room temp

suitability

bacteria (fermentation)

Other Notes

A long-lasting fatty acid ester antifoam, 100% active.

Caution

Can be autoclaved repeatedly.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The application of antifoam silicones in the microbiological fermentation technic].
L Zalay et al.
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. 1. Abt. Medizinisch-hygienische Bakteriologie, Virusforschung und Parasitologie. Originale, 197(1), 118-126 (1965-07-01)
S Zhang et al.
Journal of biotechnology, 25(3), 289-306 (1992-09-01)
Foam formation and the subsequent cell damage/losses in the foam layer were found to be the major problems affecting cell growth and monoclonal antibody (MAb) production in stirred and sparged bioreactors for both serum-supplemented and serum-free media. Surfactants in the
L A van der Pol et al.
Biotechnology progress, 9(5), 504-509 (1993-09-01)
Addition of high concentrations of silicone antifoam (AF) to a suspension of hybridoma cells in a bubble column reduces the death rate when using medium without a protective component. In contrast, high AF concentrations can cause detrimental effects in a
J R Hermann et al.
Applied and environmental microbiology, 72(7), 4811-4818 (2006-07-06)
The objective of this research was to optimize sampling parameters for increased recovery and detection of airborne porcine reproductive and respiratory syndrome virus (PRRSV) and swine influenza virus (SIV). Collection media containing antifoams, activated carbons, protectants, and ethylene glycol were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service