A7790
Albumin, α-D-mannopyranosylphenyl isothiocyanate and fluorescein isothiocyanate labeled bovine
lyophilized powder
Synonym(s):
α-D-Mannosylated-FITC-albumin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
biological source
bovine
form
lyophilized powder
extent of labeling
1.5-3.0 mol fluorescein per mol albumin
impurities
salt, free
storage temp.
−20°C
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Fertility and sterility, 70(3), 482-491 (1998-10-03)
To assess whether mannose receptor assays can predict fertilization outcome in vitro. A prospective, double-blind study of the mannose receptor properties of spermatozoa. Assisted human reproduction program at a university hospital. Partners of 140 consecutive women undergoing their first in
Archives of andrology, 38(1), 67-74 (1997-01-01)
The objective of this study was to characterize patterns of surface expression of mannose-binding sites (MBS) on human spermatozoa while evaluating the influence of sperm viability, plasma membrane integrity, and capacitation, D-Mannose binding sites were visualized by fluorescence microscopy using
Nature communications, 6, 6801-6801 (2015-04-16)
Liver receptor homologue-1 (LRH-1) is a nuclear receptor involved in the repression of inflammatory processes in the hepatointestinal tract. Here we report that LRH-1 is expressed in macrophages and induced by the Th2 cytokine IL-13 via a mechanism involving STAT6.
Journal of neuroscience research, 91(12), 1572-1580 (2013-10-10)
Complex carbohydrate structures are essential molecules of infectious bacteria, parasites, and host cells and are involved in cell signaling associated with immune responses, glycoprotein homeostasis, and cell migration. The uptake of mannose-tailed glycans is usually carried out by professional phagocytes
Andrologia, 32(4-5), 317-329 (2000-10-06)
Zinc and lead concentrations were measured in seminal plasma from fertile donors, infertile men with varicocoele and men undergoing work-ups for in vitro fertilization. Ejaculated spermatozoa from these subjects were incubated in vitro with various metal ions and/or dibromoethane and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service