Skip to Content
Merck
All Photos(1)

Key Documents

A7380

Sigma-Aldrich

L-Arginine hydroxamate hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H15N5O2 · HCl
CAS Number:
Molecular Weight:
225.68
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

−20°C

SMILES string

Cl.N[C@@H](CCCNC(N)=N)C(=O)NO

Biochem/physiol Actions

L-Arginine hydroxamate is used as a selection agent for regulatory mutants impaired in the formation of the DNA-binding protein and in which the expression of an argF::lacZ fusion is not controlled by arginine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kisumi et al.
Applied microbiology, 22(6), 987-991 (1971-12-01)
l-Arginine hydroxamate inhibited the growth of various bacteria, and the inhibition was readily reversed by arginine. l-Arginine hydroxamate (10(-3)m) completely inhibited the growth of Bacillus subtilis. This inhibitory effect was prevented by 2.5 x 10(-4)ml-arginine, which was the most effective
S M Park et al.
Journal of bacteriology, 179(17), 5300-5308 (1997-09-01)
Gel retardation experiments indicated the presence in Pseudomonas aeruginosa cell extracts of an arginine-inducible DNA-binding protein that interacts with the control regions for the car and argF operons, encoding carbamoylphosphate synthetase and anabolic ornithine carbamoyltransferase, respectively. Both enzymes are required

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service