Skip to Content
Merck
All Photos(1)

Documents

A7331

Sigma-Aldrich

trans-2-Aminocyclohexanecarboxylic acid

≥97% (titration)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
Beilstein:
3196018
MDL number:
UNSPSC Code:
12352200

Assay

≥97% (titration)

mp

~255 °C (dec.)

SMILES string

N[C@@H]1CCCC[C@H]1C(O)=O

InChI

1S/C7H13NO2/c8-6-4-2-1-3-5(6)7(9)10/h5-6H,1-4,8H2,(H,9,10)/t5-,6-/m1/s1

InChI key

USQHEVWOPJDAAX-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William C Pomerantz et al.
Journal of the American Chemical Society, 133(34), 13604-13613 (2011-08-06)
We have examined the effect of β-peptide modifications on the propensity of these helical molecules to form lyotropic liquid crystalline (LC) phases in water. All of the β-peptides we have examined contain 10 residues. In each case, at least three
D H Appella et al.
Nature, 387(6631), 381-384 (1997-05-22)
Proteins and RNA are unique among known polymers in their ability to adopt compact and well-defined folding patterns. These two biopolymers can perform complex chemical operations such as catalysis and highly selective recognition, and these functions are linked to folding
Ulf Strijowski et al.
Organic & biomolecular chemistry, 2(8), 1105-1109 (2004-04-06)
A series of cyclic peptides containing either cis- or trans-2-aminocyclohexane carboxylic acid as mimics for L-proline has been synthesized and their structural properties have been investigated using NMR and MD methods.
Anasztázia Hetényi et al.
Journal of the American Chemical Society, 127(2), 547-553 (2005-01-13)
Homo-oligomers constructed by using trans-2-aminocyclohexanecarboxylic acid monomers without protecting groups were studied. Both ab initio theory and NMR measurements showed that the tetramer tends to adopt a 10-helix motif, while the pentamer and hexamer form the known 14-helix. It was
Betas are brought into the fold.
B L Iverson
Nature, 385(6612), 113-113 (1997-01-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service