Skip to Content
Merck
All Photos(1)

Key Documents

A7185

Sigma-Aldrich

Ala-Lys(TFA)-Pro-Ser-Tyr-Hyp-Hyp-Thr-Tyr-Lys

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C57H81N12O18F3
CAS Number:
Molecular Weight:
1279.32
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(N)C(=O)NC(CCCCNC(=O)C(F)(F)F)C(=O)N1CCCC1C(=O)NC(CO)C(=O)NC(Cc2ccc(O)cc2)C(=O)N3CC(O)CC3C(=O)N4CC(O)CC4C(=O)NC(C(C)O)C(=O)NC(Cc5ccc(O)cc5)C(=O)NC(CCCCN)C(O)=O

InChI

1S/C57H81F3N12O18/c1-29(62)46(79)64-37(8-4-6-20-63-56(90)57(58,59)60)52(85)70-21-7-10-42(70)49(82)68-41(28-73)48(81)67-40(23-32-13-17-34(76)18-14-32)53(86)72-27-36(78)25-44(72)54(87)71-26-35(77)24-43(71)50(83)69-45(30(2)74)51(84)66-39(22-31-11-15-33(75)16-12-31)47(80)65-38(55(88)89)9-3-5-19-61/h11-18,29-30,35-45,73-78H,3-10,19-28,61-62H2,1-2H3,(H,63,90)(H,64,79)(H,65,80)(H,66,84)(H,67,81)(H,68,82)(H,69,83)(H,88,89)

InChI key

JSVBNGKZVWYTKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Bioadhesive peptide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swerdloff, M.D.
International Journal of Peptide and Protein Research, 33, 318-318 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service