Skip to Content
Merck
All Photos(1)

Key Documents

A5894

Sigma-Aldrich

Antipyrylazo III

~95%

Synonym(s):

3,6-Bis(4-antipyrylazo)-4,5-dihydroxy-2,7-naphthalenedisulfonic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H26N8Na2O10S2
CAS Number:
Molecular Weight:
792.71
Beilstein:
956469
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

~95%

technique(s)

titration: suitable

suitability

suitable for determination of calcium (method in Anal. Chim. Acta, 71, 343 (1974))

SMILES string

[Na+].[Na+].CN1N(c2ccccc2)C(=O)C(\N=N\c3c(O)c4c(O)c(\N=N\C5=C(C)N(C)N(c6ccccc6)C5=O)c(cc4cc3S([O-])(=O)=O)S([O-])(=O)=O)=C1C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cation complexation by antipyrylazo III: chelate spectral behavior correlated with divalent cation properties.
S E Pattison et al.
Analytical biochemistry, 247(1), 181-183 (1997-04-05)
J Maylie et al.
The Journal of physiology, 442, 551-567 (1991-10-01)
1. The effects of 2,3-butanedione monoxime (BDM) on the optical retardation and myoplasmic Ca2+ signal were studied in twitch fibres of Rana temporaria. The myoplasmic Ca2+ transient in response to action potential stimulation was monitored in cut fibres containing Antipyrylazo
A Barth et al.
Biochimica et biophysica acta, 1057(1), 115-123 (1991-03-01)
Fourier transform infrared spectroscopy was used to investigate ligand binding and conformational changes in the Ca2(+)-ATPase of sarcoplasmic reticulum during the catalytic cycle. The ATPase reaction was started in the infrared sample by release of ATP from the inactive, photolabile
A V Tepikin et al.
The Journal of membrane biology, 123(1), 43-47 (1991-07-01)
Simultaneous optical measurements of extra- and intracellular Ca2+ concentrations were carried out on isolated snail neurons injected iontophoretically with Ca2+. The fluorescent indicator Fura-2 was used to measure intracellular concentration of free Ca, and the absorbant indicator Antipyrylazo III to
H Sun et al.
Biochimica et biophysica acta, 1403(1), 57-71 (1998-06-12)
The accurate measurement of the intracellular concentration of free magnesium ions ([Mg2+]i) is essential for evaluating the role of Mg2+ in cellular functions. Among the specific (compared to fluorescent indicators) metallochromic dyes, antipyrylazo III (APIII) appears to be most suitable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service