Skip to Content
Merck
All Photos(1)

Key Documents

A5713

Sigma-Aldrich

Monoclonal Anti-polyHistidine–Agarose antibody produced in mouse

clone HIS-1, purified immunoglobulin, PBS suspension, suitable for purification of HIS tagged recombinant proteins

Synonym(s):

Monoclonal Anti-polyHistidine, Monoclonal 6 His epitope tag, Monoclonal 6xHis-tag, Monoclonal HHHHHH epitope tag, Monoclonal Hexa His tag, Monoclonal His-tag, Monoclonal His6 tag, Monoclonal Histidine tagged, Monoclonal Poly-His-tag

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$1,490.00

$1,490.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$1,490.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

$1,490.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HIS-1, monoclonal

form

PBS suspension

isotype

IgG2a

capacity

15-30 nmol/mL binding capacity

suitability

suitable for purification of HIS tagged recombinant proteins

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

This mouse IgG2a monoclonal recombinant antibody recognizes native and denatured, reduced forms of synthetic polyhistidine and polyhistidine tagged fusion proteins.

Specificity

The antibody recognizes synthetic polyHistidine, as well as native or denatured, reduced forms of proteins tagged with 6X histidines, expressed in selected vectors.

Immunogen

recombinant polyHistidine tagged fusion protein.

Application

Antibody suitable for immunoprecipitation and immunoaffinity purification of polyhistidine and polyhistidine tagged fusion proteins from bacteria, bacterial lysates, cells and tissues transfected with polyhistidine fusion protein expressing vectors
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Preparation Note

Prepared using Protein A purified antibody coupled to cyanogen bromide-activated agarose at 2.0-2.4 mg antibody per mL resin volume.

Legal Information

This Product is covered by patent DE 19507166 and foreign equivalents exclusively licensed to QIAGEN GmbH, Qiagen Strasse 1, D-40724 Hilden, Germany.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric A Davidson et al.
Current protocols in molecular biology, Chapter 24, Unit 24-Unit 24 (2009-07-04)
This unit describes a protocol for the directed evolution of proteins utilizing in vitro compartmentalization. This method uses a large number of independent in vitro transcription and translation (IVTT) reactions in water droplets suspended in an oil emulsion to enable
Ryan T Cameron et al.
Molecular and cellular neurosciences, 61, 46-55 (2014-05-27)
Up-regulation of Hsp20 protein levels in response to amyloid fibril formation is considered a key protective response against the onset of Alzheimer's disease (AD). Indeed, the physical interaction between Hsp20 and Aβ is known to prevent Aβ oligomerisation and protects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service