Skip to Content
Merck
All Photos(1)

Key Documents

A4778

Sigma-Aldrich

Adenylosuccinic acid sodium salt

~96% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18N5O11P
CAS Number:
Molecular Weight:
463.29
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

Assay

~96% (HPLC)

storage temp.

−20°C

SMILES string

[Na].OC1C(O)C(OC1COP(O)(O)=O)n2cnc3c(NC(CC(O)=O)C(O)=O)ncnc23

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service