Skip to Content
Merck
All Photos(1)

Documents

A4511

Sigma-Aldrich

5(6)-Azidofluorescein diacetate

~95% (HPLC)

Synonym(s):

Azido-FDA, Fluorescein 5(6)-azide diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H15N3O7
CAS Number:
Molecular Weight:
457.39
MDL number:
UNSPSC Code:
12000000

Assay

~95% (HPLC)

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5ccc(cc45)N=[N+]=[N-])c1

Looking for similar products? Visit Product Comparison Guide

Application

A photolabelling reagent in membrane viscosity studies.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Rotman et al.
Biochemistry, 20(21), 5995-5999 (1981-10-13)
The intracellular viscosity changes that occur in washed human platelets as a result of activation by thrombin or ADP were studied by the use of a fluorescent probe. Results obtained showed a sharp and quick decrease of the intracellular viscosity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service