Skip to Content
Merck
All Photos(1)

Key Documents

A4355

Sigma-Aldrich

Monoclonal Anti-ASPP1 antibody produced in mouse

~2 mg/mL, clone LXO54.2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Apoptosis-stimulating protein of p53, 1, Anti-PPP1R13B, Anti-protein phosphatase 1 regulatory subunit 13B

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$1,030.00

$1,030.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
$1,030.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$1,030.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LXO54.2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~175 kDa

species reactivity

mouse, human

concentration

~2 mg/mL

technique(s)

immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract of human osteogenic sarcoma, U-2-OS

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

ASPP1 interacts with p53 to enhance p53-induced apoptosis.
Monoclonal Anti-ASPP1 (mouse IgG1 isotype) is derived from the hybridoma LXO54.2 produced by the fusion of mouse myeloma cells (SP2/0 cells) and splenocytes from BALB/c mice immunized with a recombinant fragment of human ASPP1, amino acids. Apoptosis-stimulating protein of p53, 1 (ASPP1) belongs to the ASPP family of proteins. This protein contains a proline-rich region, four ankyrin repeats, and an SH3 domain in the C-terminal end.

Immunogen

recombinant fragment of human ASPP1 (amino acids 1-308).

Application

Monoclonal Anti-ASPP1 antibody can be used for western blot at 1-2 μg/mL using total cell extract of human osteogenic sarcoma, U-2-OS. The antibody can also be used for microarray and immunoprecipitation assays.

Biochem/physiol Actions

Apoptosis-stimulating protein of p53, 1 (ASPP1) is known to stimulate RAS signalling which subsequently enhances p53-dependent apoptosis in cancer cells. Furthermore, ASPP1 can also induce p53-independent apoptosis by enhancing the apoptotic functions of p63 and p73. Monoclonal Anti-ASPP1 antibody recognizes human and mouse ASPP1.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingwen Wang et al.
International journal of cancer, 141(7), 1422-1433 (2017-06-29)
Inactivation of p53 has been shown to correlate with drug resistance in tumors. However, in clear cell renal cell carcinoma (ccRCC), p53 is rarely mutated, yet the tumors remain highly insensitive to the conventional chemotherapeutic drugs. The underlying mechanisms responsible
Marcus M Schittenhelm et al.
Cell death & disease, 15(1), 25-25 (2024-01-10)
ASPP1 (PPP1R13B) belongs to a family of p53-binding proteins and enhances apoptosis by stimulation of p53-transactivation of selected proapoptotic target genes. It is preferentially expressed in hematopoietic stem cells (HSC) and together with p53 preserves the genomic integrity of the
Victor C Y Mak et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 24(4), 522-532 (2010-11-26)
Gestational trophoblastic disease encompasses a spectrum of trophoblastic lesions including true neoplasms such as choriocarcinomas and the potentially malignant hydatidiform moles, which may develop persistent disease requiring chemotherapy. ASPP1, a member of apoptosis-stimulating proteins of p53 (ASPPs), is a proapoptotic
Libin Yin et al.
Oncology letters, 15(5), 6409-6417 (2018-05-08)
The apoptosis-stimulating protein of p53 (ASPP) family is a newly identified family protein including ASPP1, ASPP2 and inhibitor of ASPP (iASPP), by which the tumor protein 53 (TP53)-mediated apoptotic process is selectively regulated. Downregulation of ASPP1/ASPP2 and upregulation of iASPP
Daniele Bergamaschi et al.
Molecular and cellular biology, 24(3), 1341-1350 (2004-01-20)
We recently showed that ASPP1 and ASPP2 stimulate the apoptotic function of p53. We show here that ASPP1 and ASPP2 also induce apoptosis independently of p53. By binding to p63 and p73 in vitro and in vivo, ASPP1 and ASPP2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service