A2641
4-Aza-DL-leucine dihydrochloride
Synonym(s):
β-Dimethylamino-DL-alanine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C5H12N2O2 · 2HCl
CAS Number:
Molecular Weight:
205.08
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥98.0% (TLC)
form
powder
color
white to off-white
storage temp.
−20°C
SMILES string
OC(C(N)CN(C)C)=O.[H]Cl.[H]Cl
InChI
1S/C5H12N2O2.ClH/c1-7(2)3-4(6)5(8)9;/h4H,3,6H2,1-2H3,(H,8,9);1H
InChI key
JXAGRLBYOYAIRY-UHFFFAOYSA-N
Biochem/physiol Actions
4-Aza-DL-leucine is used as a leucine analogue that is toxic for bacterial high-affinity branched-chain amino acid (BCAA) transport system (LIV-I). 4-Aza-DL-leucine is used to select mutants of the LIV-1 transporter.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Azusa Saika et al.
AMB Express, 1(1), 6-6 (2011-09-13)
Ralstonia eutropha PHB-4 expressing Pseudomonas sp. 61-3 polyhydroxyalkanoate (PHA) synthase 1 (PhaC1Ps) synthesizes PHA copolymer containing 3-hydroxybutyrate (3HB) and a small amount (0.5 mol%) of 3-hydroxy-4-methylvalerate (3H4MV) from fructose as a carbon source. In this study, enhanced incorporation of 3H4MV
T Hoshino et al.
Journal of bacteriology, 172(10), 5540-5543 (1990-10-01)
About 30 mutants of Pseudomonas aeruginosa PAO defective in the high-affinity branched-chain amino acid transport system (LIV-I) were isolated by the selection for resistance to 4-aza-DL-leucine, a toxic leucine analog for LIV-I. All of the mutants were complemented by plasmid
Feng Wang et al.
Journal of bacteriology, 191(8), 2574-2584 (2009-02-10)
Members of the genus Shewanella inhabit various environments; they are capable of synthesizing various types of low-melting-point fatty acids, including monounsaturated fatty acids (MUFA) and branched-chain fatty acids (BCFA) with and without eicosapentanoic acid (EPA). The genes involved in fatty
Haifeng Su et al.
Scientific reports, 6, 39543-39543 (2016-12-21)
Biosynthesis approaches for the production of higher alcohols as a source of alternative fossil fuels have garnered increasing interest recently. However, there is little information available in the literature about using undirected whole-cell mutagenesis (UWCM) in vivo to improve higher
Takashi Koyanagi et al.
FEMS microbiology letters, 237(1), 73-77 (2004-07-23)
The growth of Escherichia coli is inhibited by an antibiotic compound, azaserine (O-diazoacetyl-L-serine). Previous studies revealed the biochemical properties of azaserine, which involves inhibition of various enzymatic reactions as well as introduction of DNA breakage. However, genetically, nothing has been
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service