Skip to Content
Merck
All Photos(1)

Documents

A2297

Sigma-Aldrich

Ala-Ser-Thr-Thr-Thr-Asn-Tyr-Thr acetate salt

≥97% (HPLC)

Synonym(s):

Peptide T

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H55N9O16
CAS Number:
Molecular Weight:
857.86
UNSPSC Code:
12352200

Assay

≥97% (HPLC)

storage temp.

−20°C

Biochem/physiol Actions

HIV inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C B Pert et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(23), 9254-9258 (1986-12-01)
The differentiation antigen T4, present on the helper/inducer subset of T lymphocytes, is thought to serve as the receptor for the human immunodeficiency virus (HIV). We find that a 60-kDa protein, immunoprecipitable by monoclonal antibody (mAb) OKT4, is present on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service