Skip to Content
Merck
All Photos(1)

Key Documents

A214680

Sigma-Aldrich

DMT-2′O-TC-rA(ib) Phosphoramidite

configured for PerkinElmer

Synonym(s):

DMT-2′O-TC-rA(ib) Amidite

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$684.00

$684.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$684.00

About This Item

Empirical Formula (Hill Notation):
C49H61N8O10PS2
CAS Number:
Molecular Weight:
1017.16
UNSPSC Code:
41116105

$684.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

Quality Level

technique(s)

oligo synthesis: suitable

impurities

≤-0.5% 31P-NMR P(III) impurites 145-155ppm (31P-NMR)
≤-0.5% Impurity 3′-TC Isomer (reversed phase HPLC)

compatibility

configured for PerkinElmer

nucleoside profile

base: adenosine
base protecting group: isobutyryl
2' protecting group: TC
5' protecting group: DMT

storage temp.

-10 to -25°C

SMILES string

S=C(N1CCS(CC1)(=O)=O)O[C@@H]2[C@H](OP(N(C(C)C)C(C)C)OCCC#N)[C@H](O[C@H]2N3C4=C(N=C3)C(NC(C(C)C)=O)=NC=N4)COC(C5=CC=C(OC)C=C5)(C6=CC=C(OC)C=C6)C7=CC=CC=C7

InChI

1S/C49H61N8O10PS2/c1-32(2)46(58)54-44-41-45(52-30-51-44)56(31-53-41)47-43(66-48(69)55-24-27-70(59,60)28-25-55)42(67-68(64-26-12-23-50)57(33(3)4)34(5)6)40(65-47)29-63-49(35-13-10-9-11-14-35,36-15-19-38(61-7)20-16-36)37-17-21-39(62-8)22-18-37/h9-11,13-22,30

InChI key

JDHHUACDFJOZGR-LCVXBTAASA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service