Skip to Content
Merck
All Photos(1)

Key Documents

A1862

Sigma-Aldrich

Alpidem

≥98% (HPLC), powder

Synonym(s):

6-Chloro-2-(4-chlorophenyl)-N,N-dipropyl-imidazo[1,2-a]pyridine-3-acetamide, 6-Chloro-2-(p-chlorophenyl)-N,N-dipropylimidazo(1,2-a)pyridine-3-acetamide, Ananxyl, SL 80.0342-00

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23Cl2N3O
CAS Number:
Molecular Weight:
404.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥10 mg/mL

originator

Sanofi Aventis

storage temp.

2-8°C

SMILES string

CCCN(CCC)C(=O)Cc1c(nc2ccc(Cl)cn12)-c3ccc(Cl)cc3

InChI

1S/C21H23Cl2N3O/c1-3-11-25(12-4-2)20(27)13-18-21(15-5-7-16(22)8-6-15)24-19-10-9-17(23)14-26(18)19/h5-10,14H,3-4,11-13H2,1-2H3

InChI key

JRTIDHTUMYMPRU-UHFFFAOYSA-N

Gene Information

human ... TSPO(706)

Biochem/physiol Actions

Alpidem is a potent antagonist of peripheral benzodiazepine receptor (PBR) that is located on the outer mitochondrial membrane and interacts with the mitochondrial permeability transition (MPT) pore. Alpidem is an anxiolytic drug from the imidazopyridine family. Alpidem acts selectively on the α3 receptor subtype and to a lesser extent at the α1 subtype (Kd of 0.33nM and 1.67nM respectively), of the benzodiazepine receptor.
Alpidem is a potent peripheral benzodiazepine receptor (PBR) antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Sanofi Aventis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Chodera et al.
Polish journal of pharmacology, 46(5), 479-481 (1994-09-01)
The development of tolerance to the pharmacodynamic effects of azopirone (buspirone) and imidazopyridines (alpidem and zolpidem) was investigated. It was found that tolerance to the anxiolytic effect of buspirone develops only after 42 days of administration (twice daily). Of the
J F O'Hanlon et al.
Neuropsychobiology, 31(2), 81-88 (1995-01-01)
Effects of benzodiazepine (diazepam, lorazepam) and benzodiazepine-like anxiolytics (alpidem, suriclone) and a 5-HT-3 antagonist (ondansetron) on actual driving performance were measured in three double-blind, placebo-controlled studies. Subjects were healthy volunteers in two and anxious patients in the third. Treatments lasted
D J Sanger et al.
Psychopharmacology, 113(3-4), 395-403 (1994-01-01)
Alpidem in an imidazopyridine derivative which binds selectively to the omega 1 (BZ1) receptor subtype. It is active in some, but not all, behavioural tests sensitive to benzodiazepine anxiolytics and has clinical anti-anxiety effects. However, in a previous study, it
[Subfulminant hepatitis caused by alpidem and treated by liver transplantation].
P Ausset et al.
Gastroenterologie clinique et biologique, 19(2), 222-223 (1995-02-01)
[A new case of hepatitis after alpidem monotherapy].
B Prat et al.
Gastroenterologie clinique et biologique, 18(10), 903-904 (1994-01-01)

Articles

We offer many products related to GABAA receptors for your research needs.

DISCOVER Bioactive Small Molecules for Neuroscience

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service