Skip to Content
Merck
All Photos(1)

Documents

A1840

Millipore

Ashby′s mannitol agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Application

For the cultivation of Azotobacter that use mannitol as a source of carbon.

Components

Ingredients (g/L):
Mannitol, 20.00
Potasium phosphate dibasic, 0.20
Magnesium sulfate, 0.20
Sodium chloride, 0.20
Potassium sulfate, 0.10
Calcium carbonate, 5.00
Agar, 15.00

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service