Skip to Content
Merck
All Photos(1)

Key Documents

A182

Sigma-Aldrich

trans-(1S,3R)-ACPD

>97%, solid

Synonym(s):

trans-(1S,3R)-1-Amino-1,3-cyclopentanedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO4
CAS Number:
Molecular Weight:
173.17
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>97%

form

solid

color

white

solubility

ethanol: 0.24 mg/mL
H2O: 20 mg/mL

SMILES string

N[C@]1(CC[C@H](C1)C(O)=O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective metabotropic glutamate receptor agonist; active enantiomer of trans-(±)-ACPD.

Other Notes

Note: under IUPAC nomenclature, this compound is designated cis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W McDonald et al.
European journal of pharmacology, 215(2-3), 353-354 (1992-05-14)
The role of metabotropic type excitatory amino acid receptors in brain injury was assessed using the selective metabotropic receptor agonist, (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid (1S,3R-ACPD). Intrastriatal stereotaxic injection of 1S,3R-ACPD (250 nmol) in PND 7 rats produced little brain injury as assessed
D D Schoepp et al.
Neuroscience letters, 145(1), 100-104 (1992-09-28)
Metabotropic glutamate receptors are selectively activated by 1S,3R-1-aminocyclopentane-1,3-dicarboxylic acid (1S,3R-ACPD). [3H]Glutamate binding sites in rat brain membranes were characterized in the presence of (RS)-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA), kainate, and N-methyl-D-aspartate (NMDA) to block binding to ionotropic glutamate receptors. 1S,3R-ACPD displaced a
G N Andrianov et al.
Hearing research, 204(1-2), 200-209 (2005-06-01)
Using immunocytochemistry and multiunit recording of afferent activity of the whole vestibular nerve, we investigated the role of metabotropic glutamate receptors (mGluR) in the afferent neurotransmission in the frog semicircular canals (SCC). Group I (mGluR1alpha) and group II (mGluR2/3) mGluR
Víctor M Blanco et al.
American journal of physiology. Heart and circulatory physiology, 294(6), H2855-H2863 (2008-05-06)
A growing number of studies support an important contribution of astrocytes to neurovascular coupling, i.e., the phenomenon by which variations in neuronal activity trigger localized changes in blood flow that serve to match the metabolic demands of neurons. However, since
Matthew E Larkum et al.
Journal of neurophysiology, 99(2), 683-694 (2007-11-30)
The six-layered mammalian neocortex evolved from the three-layered paleocortex, which is retained in present-day reptiles such as the turtle. Thus the turtle offers an opportunity to examine which cellular and circuit properties are fundamental to cortical function. We characterized the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service