Skip to Content
Merck
All Photos(3)

Key Documents

A1486

Sigma-Aldrich

Anti-ATP6AP1 (451-465) antibody produced in rabbit

IgG fraction of antiserum, PBS solution

Synonym(s):

Anti-16A, Anti-ATP6IP1, Anti-ATP6S1, Anti-Ac45, Anti-CF2, Anti-VATPS1, Anti-Vacuolar ATP synthase subunit S1 precursor, Anti-XAP3

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
$766.00

$766.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
$766.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$766.00


Estimated to ship on31 May 2025


Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

PBS solution

mol wt

antigen ~52 kDa

species reactivity

human

technique(s)

western blot: 1:500-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP6AP1(537)

General description

ATPase H+ transporting accessory protein 1 (ATP6AP1) is an accessory subunit of the V-ATPase. This gene is located on human chromosome Xq28. ATP6AP1 is also referred as Ac45. It is abundantly expressed a high levels in neuronal and (neuro-) endocrine cells and osteoclasts.

Immunogen

synthetic peptide corresponding to amino acids 451-465 of human ATP6AP1

Application

Rabbit anti-ATP6AP1 (451-465) antibody can be used for western blot (1:500-1:2,000) assays.
Yale Center for High Throughput Cell Biology IF-tested antibodies. Each antibody is tested by immunofluorescence against HUVEC cells using the Yale HTCB IF protocol. To learn more about us and Yale Center for High Throughput Cell Biology partnership, visit sigma.com/htcb-if.

Biochem/physiol Actions

ATPase H+ transporting accessory protein 1 (ATP6AP1) is known to cause an X-linked N-glycosylation syndrome with liver disease. It is essential for endosomal acidification. It also participates in membrane trafficking and Ca2+-dependent membrane fusion.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expanding the phenotype of metabolic cutis laxa with an additional disorder of N-linked protein glycosylation
Witters P, et al.
European Journal of Human Genetics, 26(5), 618-618 (2018)
Loss-of-function mutations in ATP6AP1 and ATP6AP2 in granular cell tumors
Pareja F, et al.
Nature Communications, 9(1), 3533-3533 (2018)
Jason J Gokey et al.
Developmental biology, 407(1), 115-130 (2015-08-09)
Asymmetric fluid flows generated by motile cilia in a transient 'organ of asymmetry' are involved in establishing the left-right (LR) body axis during embryonic development. The vacuolar-type H(+)-ATPase (V-ATPase) proton pump has been identified as an early factor in the
Fresia Pareja et al.
Nature communications, 9(1), 3533-3533 (2018-09-01)
Granular cell tumors (GCTs) are rare tumors that can arise in multiple anatomical locations, and are characterized by abundant intracytoplasmic granules. The genetic drivers of GCTs are currently unknown. Here, we apply whole-exome sequencing and targeted sequencing analysis to reveal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service