Skip to Content
Merck
All Photos(1)

Key Documents

A1108

Sigma-Aldrich

ALK4 (150-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ACTRIB, ACVR1 B, ACVRLK4, SKR2

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$537.00

$537.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
10 μG
$537.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$537.00


Estimated to ship on24 April 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

14-18 nmol/min·mg

mol wt

~64 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ACVR1B(91)

Biochem/physiol Actions

ALK4 is a member of the subfamily of receptor ser/thr kinases that mediates signaling by the Activins. ALK4 is expressed in many human tissues, including kidney, pancreas, brain, lung, and liver. Truncated ALK4, predominantly expressed in human pituitary adenomas, function as dominant negative receptors to interfere with wild-type receptor function and blocks the antiproliferative effect of activin possibly contributing to development of human pituitary tumors. ALK4 is able to mediate Nodal signaling in the presence of Cripto during vertebrate development.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Reissmann et al.
Genes & development, 15(15), 2010-2022 (2001-08-04)
Nodal proteins have crucial roles in mesendoderm formation and left-right patterning during vertebrate development. The molecular mechanisms of signal transduction by Nodal and related ligands, however, are not fully understood. In this paper, we present biochemical and functional evidence that
Y Zhou et al.
Molecular endocrinology (Baltimore, Md.), 14(12), 2066-2075 (2000-12-16)
Activin, a member of the transforming growth factor beta (TGFbeta) superfamily of cytokines, inhibits cell proliferation in a variety of cell types. The functions of activin are mediated by type I and type II serine/threonine kinase receptors. The main type

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service