94333
Uridine 5′-diphosphogalactose disodium salt
≥98.0% (HPLC)
Synonym(s):
UDP-Gal, Uridine-diphosphate-galactose disodium salt, Uridine[5’]diphospho[1]-α-D-galactopyranose disodium salt
About This Item
Recommended Products
Assay
≥98.0% (HPLC)
form
powder
impurities
≤5% solvent (ethanol)
≤6% water
solubility
H2O: 50 mg/mL, clear, colorless
storage temp.
−20°C
SMILES string
[Na+].[Na+].OC[C@H]1O[C@@H](OP([O-])(=O)OP([O-])(=O)OC[C@H]2OC([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O)[C@H](O)[C@@H](O)[C@H]1O
InChI
1S/C15H24N2O17P2.2Na/c18-3-5-8(20)10(22)12(24)14(32-5)33-36(28,29)34-35(26,27)30-4-6-9(21)11(23)13(31-6)17-2-1-7(19)16-15(17)25;;/h1-2,5-6,8-14,18,20-24H,3-4H2,(H,26,27)(H,28,29)(H,16,19,25);;/q;2*+1/p-2/t5-,6-,8+,9-,10+,11-,12-,13?,14?;;/m1../s1
InChI key
PKJQEQVCYGYYMM-OUJOOSCPSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service