Skip to Content
Merck
All Photos(1)

Key Documents

93759

Sigma-Aldrich

Pyridoxal hydrochloride

≥99.5% (T)

Synonym(s):

3-Hydroxy-5-(hydroxymethyl)-2-methyl-4-pyridinecarboxaldehyde hydrochloride, PL HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO3 · HCl
CAS Number:
Molecular Weight:
203.62
Beilstein:
3656994
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥99.5% (T)

form

powder

loss

≤0.6% loss on drying

color

white to faint beige

mp

173 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤1000 mg/kg

cation traces

Ca: ≤250 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤15 mg/kg
Cu: ≤5 mg/kg
Fe: ≤15 mg/L
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

cell analysis

SMILES string

Cl[H].[H]C(=O)c1c(CO)cnc(C)c1O

InChI

1S/C8H9NO3.ClH/c1-5-8(12)7(4-11)6(3-10)2-9-5;/h2,4,10,12H,3H2,1H3;1H

InChI key

FCHXJFJNDJXENQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridoxal is a derivative of vitamin B6.[1] Vitamin B6 derivatives contain a core pyridine ring. Pyridoxal consists of an aldehyde group at pyridine′s 4′ position.[2] Pyridoxal is a water-soluble vitamin.[3]

Application

For the labeling of amino acids and their detection in picomolar amounts; Coenzymes and Cofactors, vol. 1: Vitamin B6, Pyridoxal Phosphate

Biochem/physiol Actions

Vitamin B6 derivatives are initially oxidized to pyridoxal. It is then phosphorylated to pyridoxal 5′- phosphate (P5P). Pyridoxal 5′- phosphate is the circulatory form of vitamin B6.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of water-soluble vitamins in infant milk by high-performance liquid chromatography
Albala S, et al.
Journal of Chromatography A, 778(1-2), 247-253 (1997)
Vitamin B6 (Pyridoxine; Pyridoxal 5'-Phosphate)
CH, C
Alternative Medicine Review, 6(1), 87-87 (2001)
Thomas Bachmann et al.
Analytical and bioanalytical chemistry, 412(26), 7237-7252 (2020-08-17)
Vitamin B6 comprises an important set of molecules tightly interwoven with the human amino acid, fatty acid, and carbohydrate metabolism. Analytical methods striving for the quantification of individual B6 vitamers so far mostly rely on methods based on HPLC in
Vitamin B6 and its role in cell metabolism and physiology
Parra M, et al.
Cells, 7(7), 84-84 (2018)
Toshio Iwasaki et al.
Journal of the American Chemical Society, 131(38), 13659-13667 (2009-09-10)
CW EPR spectra of reduced [2Fe-2S](Cys)(3)(His)(1) clusters of mammalian mitoNEET soluble domain appear to produce features resulting from the interaction of the electron spins of the two adjacent clusters, which can be explained by employing the local spin model. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service