Skip to Content
Merck
All Photos(1)

Key Documents

91709

Sigma-Aldrich

Trifluoroacetic acid solution

BioReagent, suitable for protein sequencing, ~25% in H2O

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
5446934
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

product line

BioReagent

analyte chemical class(es)

amino acids

concentration

~25% in H2O

suitability

suitable for protein sequencing

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Special 40 ml packaging for direct use in common sequencers.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yukiko Enomoto-Rogers et al.
Carbohydrate polymers, 92(2), 1827-1834 (2013-02-13)
Fully substituted glucomannan triacetate (GMTAc) (degree of substitution (DS)=3.0) was prepared from konjac glucomannan (KGM) treated with acetic acid and trifluoroacetic anhydride (TFAA). The peaks in the (1)H- and (13)C NMR spectra of GMTAc were assigned in detail based on
Guillaume Plourde et al.
Lancet (London, England), 386(10009), 2192-2203 (2015-09-29)
Transradial access for cardiac catheterisation results in lower bleeding and vascular complications than the traditional transfemoral access route. However, the increased radiation exposure potentially associated with transradial access is a possible drawback of this method. Whether transradial access is associated
Jennifer Lee et al.
PloS one, 10(6), e0131506-e0131506 (2015-06-27)
Although inactivating frameshift mutations in the Transforming growth factor beta receptor type 2 (TGFBR2) gene are considered as drivers of microsatellite unstable (MSI) colorectal tumorigenesis, consequential alterations of the downstream target proteome are not resolved completely. Applying a click-it chemistry
Wolfgang Klein et al.
PloS one, 10(3), e0120886-e0120886 (2015-03-26)
The cyclodepsipeptide cotransin was described to inhibit the biosynthesis of a small subset of proteins by a signal sequence-discriminatory mechanism at the Sec61 protein-conducting channel. However, it was not clear how selective cotransin is, i.e. how many proteins are sensitive.
Rushd Khalaf et al.
Journal of chromatography. A, 1397, 11-18 (2015-05-03)
The purification of therapeutic peptides is most often performed using one or more reversed phase chromatography steps. This ensures high purities while keeping the costs of purification under control. In this paper, a doped reversed phase chromatographic material is tested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service