Skip to Content
Merck
All Photos(1)

Documents

90446

Sigma-Aldrich

Triethylene glycol monodecyl ether

BioXtra, ≥97.0% (GC)

Synonym(s):

C10E3, Decyl triethylene glycol ether, Decyltriglycol, Polyoxyethylene (3) decyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9(OCH2CH2)3OH
CAS Number:
Molecular Weight:
290.44
Beilstein:
1908434
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

product line

BioXtra

Assay

≥97.0% (GC)

form

liquid

mol wt

290.44 g/mol

refractive index

n20/D 1.449

density

0.936 g/mL at 20 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCOCCOCCOCCO

InChI

1S/C16H34O4/c1-2-3-4-5-6-7-8-9-11-18-13-15-20-16-14-19-12-10-17/h17H,2-16H2,1H3

InChI key

UKODLHVFJRCQME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Non-ionic surfactant. Has been used to study the sodium current of the squid giant axon; Effectively releases histamine from the cell granules of rat peritoneal mast cells

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

André Pierre Schroder et al.
Scientific reports, 9(1), 2292-2292 (2019-02-21)
The lamellar-to-sponge phase transition of fluorescently labelled large unilamellar vesicles (LUVs) of the non-ionic surfactant triethylene glycol mono n-decyl ether (C10E3) was investigated in situ by confocal laser scanning microscopy (CLSM). Stable dispersions of micrometer-sized C10E3 LUVs were prepared at
The effect of surfactants upon rat peritoneal mast cells in vitro.
C Prottey et al.
Food and cosmetics toxicology, 14(5), 425-430 (1976-10-01)
D A Haydon et al.
The Journal of physiology, 341, 411-427 (1983-08-01)
The effects of several n-alkanols and n-alkyl oxyethylene alcohols, methyl octanoate, glycerol 1-monooctanoate and dioctanoyl phosphatidylcholine on the ionic currents and electrical capacity of the squid giant axon membrane have been examined. The peak inward current in voltage-clamped axons was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service