89858
1-Pyrenylmethyl methanethiosulfonate
for fluorescence
Synonym(s):
Methanesulfonothioic acid S-(1-pyrenylmethyl) ester
About This Item
grade
for fluorescence
fluorescence
λex 374 nm; λem 398 nm in 0.1 M phosphate pH 7.0
suitability
corresponds for coupling to thiols
storage temp.
−20°C
SMILES string
CS(=O)(=O)SCc1ccc2ccc3cccc4ccc1c2c34
InChI
1S/C18H14O2S2/c1-22(19,20)21-11-15-8-7-14-6-5-12-3-2-4-13-9-10-16(15)18(14)17(12)13/h2-10H,11H2,1H3
InChI key
OXKQGBUJTFUGBB-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service