Skip to Content
Merck
All Photos(1)

Key Documents

84432

Sigma-Aldrich

Hydrochloric acid – propionic acid solution

1:1 (v/v), for protein sequence analysis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
grade:
for protein sequence analysis
Pricing and availability is not currently available.

grade

for protein sequence analysis

Assay

≥44% (with NaOH; CH3CH2COOH, titration)

form

liquid

quality

ampule

SMILES string

Cl.CCC(O)=O

InChI

1S/C3H6O2.ClH/c1-2-3(4)5;/h2H2,1H3,(H,4,5);1H

InChI key

YYFIGOPUHPDIBO-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of amino acids on Merrifield resin by microwave hydrolysis.
H M Yu et al.
Journal of chromatography, 456(2), 357-362 (1988-12-16)
The use of propionic acid-hydrochloric acid hydrolysis in Merrifield solid-phase peptide synthesis.
F C Westall et al.
The Journal of organic chemistry, 37(21), 3363-3365 (1972-10-20)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service