Skip to Content
Merck
All Photos(1)

Documents

77636

Sigma-Aldrich

Phenolphthalein β-D-glucuronide

≥98% (TLC)

Synonym(s):

Phenolphthalein β-D-glucosiduronic acid, Phenolphthalein glucuronic acid, Phenolphthalein mono-β-D-glucosiduronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22O10
CAS Number:
Molecular Weight:
494.45
Beilstein:
68526
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

biological source

rabbit urine

Assay

≥98% (TLC)

form

powder

impurities

≤0.5% free phenolphthalein
≤6% water

solubility

H2O: 0.1 g/mL, clear, light yellow

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(O)cc5

InChI

1S/C26H22O10/c27-15-9-5-13(6-10-15)26(18-4-2-1-3-17(18)24(33)36-26)14-7-11-16(12-8-14)34-25-21(30)19(28)20(29)22(35-25)23(31)32/h1-12,19-22,25,27-30H,(H,31,32)/t19-,20-,21+,22-,25+,26?/m0/s1

InChI key

FXJYOZKDDSONLX-XADSOVDISA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-glucuronidase test: suitable as substrate, for liver and bacterial enzymes

Other Notes

Characterization of β-glucuronidase from Ampullaria

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

T. Tsukada et al.
Comparative Biochemistry and Physiology, 86B, 565-565 (1987)
Adam G Staines et al.
Analytical biochemistry, 342(1), 45-52 (2005-06-17)
A primary reason for poor characterization of microsomal transport to date is the limitations of the measurement techniques used. Radiodetection provides sufficient sensitivity, but it can be applied only when labeled analogue is available. In this article, we report the
M Csala et al.
Biochemical pharmacology, 59(7), 801-805 (2000-03-16)
The physiological function of microsomal beta-glucuronidase is unclear. Substrates may be either glucuronides produced in the lumen of endoplasmic reticulum (ER) or those taken up by hepatocytes. In the latter case, efficient inward transport of glucuronides at the plasma membrane
Takahiro Sasamoto et al.
Journal of gastroenterology and hepatology, 21(1 Pt 1), 26-31 (2006-05-19)
There are several reports on the biliary excretion of bile acids and organic anions in zone 1- and zone 3-injured rat liver, but the results are controversial. In order to dissolve the discrepancy between previous works about the role of
J A Wilhelm et al.
Journal of chromatography, 578(2), 231-238 (1992-07-24)
A procedure is described to simultaneously quantitate phenolphthalein and its glucuronide metabolite from dog serum, urine and bile using high-performance liquid chromatography. The major advantages of this over pre-existing methods include direct analysis of the parent compound and glucuronide metabolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service