Skip to Content
Merck
All Photos(1)

Key Documents

77017

Sigma-Aldrich

1,2-Dierucoyl-sn-glycero-3-phosphocholine

≥98.0% (TLC)

Synonym(s):

PC, (7R,22Z)-4-Hydroxy-N,N,N-trimethyl-10-oxo-7-[[(13Z)-1-oxo-13-docosen-1-yl]oxy]-3,5,9-trioxa-4-phosphahentriacont-22-en-1-aminium 4-oxide, inner salt, 1,2-Di(cis-13-docosenoyl)-sn-glycero-3-phosphocholine, DEPC, PC(22:1(13E)/22:1(13E)), PC(22:1/22:1)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H100NO8P
CAS Number:
Molecular Weight:
898.33
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.85

Assay

≥98.0% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCC/C=C\CCCCCCCC)=O)=O

InChI

1S/C52H100NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-42-44-51(54)58-48-50(49-60-62(56,57)59-47-46-53(3,4)5)61-52(55)45-43-41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h20-23,50H,6-19,24-49H2,1-5H3/b22-20-,23-21-/t50-/m1/s1

InChI key

SDEURMLKLAEUAY-JFSPZUDSSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yi-Ting Sun et al.
FEBS open bio, 5, 515-521 (2015-07-15)
Antibiotic drug resistance is a serious issue for the treatment of bacterial infection. Understanding the resistance to antibiotics is a key issue for developing new drugs. We used penicillin and sulbactam as model antibiotics to study their interaction with model
Juan H Bolivar et al.
Biochimica et biophysica acta, 1858(6), 1278-1287 (2016-03-02)
Information about lipid-protein interactions for G protein-coupled receptors (GPCRs) is scarce. Here, we use electron spin resonance (ESR) and spin-labelled lipids to study lipid interactions with the rat neurotensin receptor 1 (NTS1). A fusion protein containing rat NTS1 fully able
John C Mathai et al.
The Journal of general physiology, 131(1), 69-76 (2008-01-02)
Despite intense study over many years, the mechanisms by which water and small nonelectrolytes cross lipid bilayers remain unclear. While prior studies of permeability through membranes have focused on solute characteristics, such as size, polarity, and partition coefficient in hydrophobic
Ruchi Kapoor et al.
The Journal of general physiology, 151(3), 342-356 (2019-02-24)
The two major classes of antidepressants, tricyclic antidepressants (TCAs) and selective serotonin reuptake inhibitors (SSRIs), inhibit neurotransmitter reuptake at synapses. They also have off-target effects on proteins other than neurotransmitter transporters, which may contribute to both desired changes in brain
Tatyana I Rokitskaya et al.
Biophysical journal, 115(3), 514-521 (2018-07-23)
Voltage-dependent translocation of a series of cationic rhodamine B derivatives differing in n-alkyl chain length (ethyl, butyl, octyl, dodecyl, octadecyl) from one lipid monolayer to another was studied by measuring electrical current relaxation after a voltage jump on a planar

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service