Skip to Content
Merck
All Photos(1)

Documents

56436C

SAFC

Dulbecco′s Modified Eagle′s Medium/High Modified

with 4500 mg/L dextrose, with 4.0 mM L-glutamine, without phenol red, without sodium bicarbonate, without sodium pyruvate, powder, suitable for cell culture

Pharma Manufacturing

Synonym(s):

Dulbecco’s Modified Eagle’s Medium - high glucose, DME, DMEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.71

description

for research or for further manufacturing use

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

phenol red: no
L-glutamine: yes
glucose: high
sodium pyruvate: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of glucose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Each of these media offers a different combination of L-glutamine and sodium pyruvate. Additionally, the glucose levels have been raised to 4500 mg/L, contributing to the name "DMEM/High".
Dulbecco′s Modified Eagle′s Medium (DMEM) is a modification of Basal Medium Eagle (BME) that contains four-fold concentrations of the amino acids and vitamins. The original formulation contained 1000 mg/L of dextrose and was used to culture embryonic mouse cells. Since then, it has been modified in several ways to support primary cultures of mouse and chicken cells, as well as a variety of normal and transformed cells. Additionally, the dextrose level has been raised to 4500 mg/L, contributing to the name "DMEM/High."

Quantity

Formulated to contain 13.4 grams of powder per liter of medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service