55962
Lithium 6-phospho-D-galactonate
≥80% (TLC)
Synonym(s):
6-Phospho-D-galactonic acid lithium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H13O10P · xLi+
CAS Number:
Molecular Weight:
276.14 (free acid basis)
UNSPSC Code:
12352201
NACRES:
NA.25
Recommended Products
Assay
≥80% (TLC)
form
powder
storage temp.
−20°C
Biochem/physiol Actions
Metabolite of the pentose phosphate pathway. Inhibits the T. brueci and sheep 6-phospho-gluconate dehydrogenase-catalysed NADP+-dependent oxidative decarboxylation of 6-phospho-D-gluconate (6PG) to D-ribulose 5-phosphate (Ru5P), inhibits the key glycolytic enzyme phospho-glucose isomerase. Metabolism of galactose-6-phosphate in rat heart and brain.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sugar derivatives as new 6-phosphogluconate dehydrogenase inhibitors selective for the parasite Trypanosoma brucei.
Pasti, C., et al.
Bioorganic & Medicinal Chemistry, 7, 1207-1214 (2003)
Studies on the metabolism of galactose-6-phosphate in rat heart and brain.
W D Musick et al.
Archives of biochemistry and biophysics, 168(1), 302-308 (1975-05-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service