Skip to Content
Merck
All Photos(1)

Key Documents

53833

Sigma-Aldrich

BioStab PCR optimizer (II)

Synonym(s):

PCR optimizer (II) BioStab

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$250.00

$250.00


Available to ship on11 April 2025Details



Select a Size

Change View
5 ML
$250.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

$250.00


Available to ship on11 April 2025Details


form

liquid

impurities

DNases, none detected
RNases, none detected
proteases, none detected

storage temp.

2-8°C

General description

BioStab PCR Optimizer is a PCR additive improving the yield and specifity of PCR reactions, enabling the amplification of targets difficult to achieve (fragments containing >70% GC). It allows the use of a wide range of annealing temperatures and eliminates the need for an optimization of amplification conditions.
The 5× solution delivered can be used at a 1-2× concentration in PCR.

Legal Information

Purchase of this product is accompanied by a limited license for use in the Polymerase Chain Reaction (PCR) process for research purposes only and in conjunction with a thermal cycler whose use in the automated performance of the PCR process is covered by an up-front license fee, either by payment to Applied Biosystems or as purchased, i.e., an authorized thermal cycler.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service