Skip to Content
Merck
All Photos(1)

Key Documents

52427

Sigma-Aldrich

Butyl 4-nitrophenyl hexylphosphonate

≥95.0% (HPLC)

Synonym(s):

Hexylphosphonic acid butyl 4-nitrophenyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$476.00

$476.00


Estimated to ship on24 May 2025



Select a Size

Change View
10 MG
$476.00

About This Item

Empirical Formula (Hill Notation):
C16H26NO5P
CAS Number:
Molecular Weight:
343.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$476.00


Estimated to ship on24 May 2025


Quality Level

Assay

≥95.0% (HPLC)

storage temp.

−20°C

SMILES string

O=P(OCCCC)(CCCCCC)OC1=CC=C([N+]([O-])=O)C=C1

InChI

1S/C16H26NO5P/c1-3-5-7-8-14-23(20,21-13-6-4-2)22-16-11-9-15(10-12-16)17(18)19/h9-12H,3-8,13-14H2,1-2H3

InChI key

DNVDUMRDCABCRP-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Zandonella et al.
European journal of biochemistry, 262(1), 63-69 (1999-05-07)
Fluorescent triacylglycerol analogs were synthesized as covalent inhibitors of lipase activity. The respective 1(3), 2-O-dialkylglycero-3(1)-alkyl-phosphonic acid p-nitrophenyl esters contain a fluorescent pyrenealkyl chain and a long-chain alkyl residue bound to the sn-2 and sn-1(3) positions of glycerol, respectively. The phosphonic
Racemase activity of B. cepacia lipase leads to dual-function asymmetric dynamic kinetic resolution of α-aminonitriles.
Pornrapee Vongvilai et al.
Angewandte Chemie (International ed. in English), 50(29), 6592-6595 (2011-06-03)
Nobuhiko Tokuriki et al.
Nature communications, 3, 1257-1257 (2012-12-06)
Optimization processes, such as evolution, are constrained by diminishing returns-the closer the optimum, the smaller the benefit per mutation, and by tradeoffs-improvement of one property at the cost of others. However, the magnitude and molecular basis of these parameters, and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service