Skip to Content
Merck
All Photos(2)

Key Documents

52001-027

Sigma-Aldrich

DNAstable® Blood LD

10 mL bottle, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

grade

for molecular biology

form

dry

storage temp.

room temp

Packaging

1 bottle

Physical form

10 mL screw cap bottle, DNAstable Blood LD Handbook

Legal Information

DNAstable is a registered trademark of Biomatrica

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service