Skip to Content
Merck
All Photos(1)

Documents

51731

Sigma-Aldrich

Polysucrose 70-fluorescein isothiocyanate conjugate

average mol wt 60,000-80,000

Synonym(s):

FITC-Polysucrose 70, Fluorescein isothiocyanate-Polysucrose 70 conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

mol wt

average mol wt 60,000-80,000

fluorescence

λex 498 nm; λem 521 nm in 0.1 M phosphate pH 7.0

Application

Ficoll 70-fluorescein isothiocyanate conjugate (FITC-Ficoll 70) is used to measure tissue permeability parameters such as glomerular sieving coefficients.

Analysis Note

degree of substitution 0.003-0.015

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transient and sustained increases in glomerular permeability following ANP infusion in rats.
Axelsson J, Rippe A, Rippe B.
American Journal of Physiology: Renal Physiology, 300, F24-F30 (2011)
Loss of size selectivity of the glomerular filtration barrier in rats following laparotomy and muscle trauma.
Axelsson J, Mahmutovic I, Rippe A, Rippe B.
American Journal of Physiology: Renal Physiology, 297, F577-F582 (2009)
Rapid, dynamic changes in glomerular permeability to macromolecules during systemic angiotensin II (ANG II) infusion in rats.
Axelsson J, Rippe A, Oberg CM, Rippe B.
American Journal of Physiology: Renal Physiology, 303, F790-F799 (2012)

Articles

Explore FITC-labelled polysaccharides and their properties, including impact of molecular weight on molecule dimensions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service