Skip to Content
Merck
All Photos(1)

Documents

50722

Sigma-Aldrich

Ammonium sulfate 1 M solution

Synonym(s):

Ammonium sulfate solution, Additive Screening Solution 12/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
38030408
PubChem Substance ID:

density

1.07 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Aqueous ammonium sulfate solution equivalent to indicated ammonia levels

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Ishwarya et al.
Preparative biochemistry & biotechnology, 43(1), 95-107 (2012-12-12)
A serine protease was purified 6.9-fold from the leaves of Thespesia populnea using ammonium sulfate fractionation followed by CM-cellulose and Sephadex G-100 chromatography. The purified enzyme was named populnein and was characterized. It was made up of a single polypeptide
Simon Gareth Broadley et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1450-1459 (2012-10-24)
MshB, a zinc-based deacetylase, catalyses a step in the mycothiol biosynthetic pathway that involves the deacetylation of 1-O-(2-acetamido-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcNAc-Ins), via cleavage of an amide bond, to 1-O-(2-amino-2-deoxy-α-D-glucopyranosyl)-D-myo-inositol (GlcN-Ins) and acetate. In this study, MshB was expressed, purified and crystallized. A
Urh Černigoj et al.
Journal of chromatography. A, 1281, 87-93 (2013-02-13)
To exploit different chromatographic modes for efficient plasmid DNA (pDNA) purification a novel monolithic chromatographic support bearing multimodal histamine (HISA) groups was developed and characterized. Electrostatic charge of HISA groups depends on the pH of the mobile phase, being neutral
Soo Chan Lee et al.
Eukaryotic cell, 11(11), 1391-1398 (2012-09-25)
Cryptococcus neoformans is a human-pathogenic basidiomycete that commonly infects HIV/AIDS patients to cause meningoencephalitis (7, 19). C. neoformans grows as a budding yeast during vegetative growth or as hyphae during sexual reproduction. Pseudohyphal growth of C. neoformans has been observed
Mior Ahmad Khushairi Mohd Zahari et al.
Journal of biomedicine & biotechnology, 2012, 125865-125865 (2012-11-08)
Factors influencing poly(3-hydroxybutyrate) P(3HB) production by Cupriavidus necator CCUG52238(T) utilizing oil palm frond (OPF) juice were clarified in this study. Effects of initial medium pH, agitation speed, and ammonium sulfate (NH(4))(2)SO(4) concentration on the production of P(3HB) were investigated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service