49927
Glycerol-Gelatine
for mounting (histochemical slides)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
grade
for mounting (histochemical slides)
composition
gelatin from porcine skin, 88.7 g/L
glycerol, 554 mL/L
phenol, 10.8 g/L
fluorescence
λex 200-600; λem 300-800 nm (corresponds)
storage temp.
2-8°C
SMILES string
OCC(O)CO
InChI
1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2
InChI key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
replaced by
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Qian Han et al.
Journal of medicinal chemistry, 52(9), 2786-2793 (2009-04-03)
Human kynurenine aminotransferase I (hKAT I) catalyzes the formation of kynurenic acid, a neuroactive compound. Here, we report three high-resolution crystal structures (1.50-1.55 A) of hKAT I that are in complex with glycerol and each of two inhibitors of hKAT
Naomi L Kruhlak et al.
Toxicology mechanisms and methods, 18(2-3), 217-227 (2008-01-01)
ABSTRACT Drug-induced phospholipidosis (PL) is a condition characterized by the accumulation of phospholipids and drug in lysosomes, and is found in a variety of tissue types. PL is frequently manifested in preclinical studies and may delay or prevent the development
Denis Fourches et al.
Chemical research in toxicology, 23(1), 171-183 (2009-12-18)
Drug-induced liver injury is one of the main causes of drug attrition. The ability to predict the liver effects of drug candidates from their chemical structures is critical to help guide experimental drug discovery projects toward safer medicines. In this
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service