Skip to Content
Merck
All Photos(1)

Key Documents

49122

Sigma-Aldrich

Glucopone 600 CS UP solution

~50% in H2O, paste

Synonym(s):

Alkyl-polyglucoside solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$219.00

$219.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$219.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.25

$219.00


Check Cart for Availability

Request a Bulk Order

description

non-ionic

Quality Level

form

paste

concentration

~50% in H2O

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the storage condition for the product 49122?

    1 answer
    1. Product 49122 needs to be stored in a tightly closed Dry place.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service