Skip to Content
Merck
All Photos(1)

Documents

48165

Sigma-Aldrich

Chitosan from crab shells

highly viscous

Synonym(s):

2-Amino-2-deoxy-(1→4)-β-D-glucopyranan, Poly-(1,4-β-D-glucopyranosamine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

(crab)

form

powder

impurities

≤1% insoluble matter

ign. residue

≤2% (as SO4)

loss

≤12% loss on drying

color

yellow

viscosity

>400 mPa.s, 1 % in acetic acid(20 °C)

InChI

1S/C56H103N9O39/c1-87-56(86)65-28-38(84)46(19(10-74)96-55(28)104-45-18(9-73)95-49(27(64)37(45)83)97-39-12(3-67)88-47(85)20(57)31(39)77)103-54-26(63)36(82)44(17(8-72)94-54)102-53-25(62)35(81)43(16(7-71)93-53)101-52-24(61)34(80)42(15(6-70)92-52)100-51-23(60)33(79)41(14(5-69)91-51)99-50-22(59)32(78)40(13(4-68)90-50)98-48-21(58)30(76)29(75)11(2-66)89-48/h11-55,66-85H,2-10,57-64H2,1H3,(H,65,86)/t11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48+,49+,50+,51+,52+,53+,54+,55+/m1/s1

InChI key

FLASNYPZGWUPSU-SICDJOISSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chitosan is a linear amino polysaccharide composed of approximately 20% β1,4-linked N-acetyl-D-glucosamine (GlcNAc) and approximately 80% β1,4-linked D-glucosamine (GlcN) that is prepared by the partial deacetylation of chitin in hot alkali.

Application

Chitosan is a biocompatible, antibacterial, and biodegradable polyelectrolyte with a variety of industrial and biomedical applications. Its chemical properties and low toxicity make it particularly suitable as a component of drug and gene delivery systems, and for the development of biodegradable films and scaffolds for tissue engineering.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyriaki G Zinoviadou et al.
Food & function, 3(3), 312-319 (2012-02-03)
Oil-in-water emulsions (10% w/w n-tetradecane) were prepared at pH = 5.7 by using, as surface active agents, electrostatically formed complexes of sodium stearoyl lactylate (SSL) at a concentration of 0.4% (w/w) and chitosan (CH) in a concentration range between 0
Rushita Shah et al.
Polymers, 11(12) (2019-12-19)
The application of polymeric biomaterial scaffolds utilizing crosslinking strategy has become an effective approach in these days. In the present study, the development and characterization of collagen-chitosan hydrogel film has been reported on using dual crosslinking agent's, i.e., tannic acid
Alberto Busilacchi et al.
Carbohydrate polymers, 98(1), 665-676 (2013-08-31)
The idea of using chitosan as a functional delivery aid to support simultaneously PRP, stem cells and growth factors (GF) is associated with the intention to use morphogenic biomaterials to modulate the natural healing sequence in bone and other tissues.
Liming Hu et al.
Nanoscale, 5(8), 3103-3111 (2013-03-22)
Within the past few years, chitosan-based drug delivery vehicles have become some of the most attractive to be studied. In contrast to all other polysaccharides, chitosan has demonstrated its unique characteristics for drug delivery platforms, including its active primary amino
Electrochemical biosensor applications of polysaccharides chitin and chitosan.
Wipa Suginta et al.
Chemical reviews, 113(7), 5458-5479 (2013-04-06)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service