Skip to Content
Merck
All Photos(1)

Key Documents

44365

Sigma-Aldrich

Baysilone® oil

viscosity ~1000 mPa.s,   neat(25 °C)

Synonym(s):

α-(Trimethylsilyl)-ω-(trimethylsilyloxy) polydimethylsiloxane, Silicon oil Baysilone®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211

form

liquid

viscosity

~1000 mPa.s, neat(25 °C)

lipid type

oils

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

100% polydimethylsiloxane with high and low temperature stability, good dielectric properties, low interfacial energy and chemical inertness

Legal Information

Baysilone is a registered trademark of Bayer AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service