Skip to Content
Merck
All Photos(1)

Key Documents

43951

Sigma-Aldrich

Kaempferol 3-β-D-glucopyranoside

≥97.0% (HPLC)

Synonym(s):

3,4′,5,7-Tetrahydroxyflavone 3-glucoside, 3-(β-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3-Glucosylkaempferol, Astragalin, Kaempferol 3-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97.0% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OC2=C(Oc3cc(O)cc(O)c3C2=O)c4ccc(O)cc4)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C21H20O11/c22-7-13-15(26)17(28)18(29)21(31-13)32-20-16(27)14-11(25)5-10(24)6-12(14)30-19(20)8-1-3-9(23)4-2-8/h1-6,13,15,17-18,21-26,28-29H,7H2/t13-,15-,17+,18-,21+/m1/s1

InChI key

JPUKWEQWGBDDQB-QSOFNFLRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Similar kaempferol glycosides reduce food intake and corresponding weight gain in Wistar rats. The glucoside was not tested.
Similar kaempferol glycosides reduce food intake and corresponding weight gain in Wistar rats.The glucoside was not tested.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianghui He et al.
Planta medica, 77(17), 1950-1957 (2011-06-16)
Cuscuta chinensis and Cuscuta australis, the two botanical sources of the Chinese herbal medicine Tu-Si-Zi, were distinguished from each other based on qualitative and quantitative chemical analysis. By HPLC‑DAD‑MS, a total of 36 compounds were characterized from these two Cuscuta
Min Ke et al.
Bio-medical materials and engineering, 22(1-3), 113-119 (2012-07-07)
Diabetic retinopathy (DR) is a severe complication of diabetes mellitus (DM) and often causes vision loss or even blindness. Vascular endothelial growth factor (VEGF) in the retina, which is mainly derived from Müller cells, is a crucial biological factor in
Hyang-Bok Lee et al.
Journal of the science of food and agriculture, 91(13), 2315-2321 (2011-05-14)
The application of tea seed extract (TSE) has been widely investigated because of its biological activities. In this paper, two flavonol triglycosides in TSE-camelliaside A (CamA) and camelliaside B (CamB)-were subjected to hydrolysis in the presence of two commercial enzyme
Krystyna Skalicka-Woźniak et al.
Journal of AOAC International, 94(1), 17-21 (2011-03-12)
Soxhlet extraction, ultrasound extraction, and accelerated solvent extraction (ASE), followed by RP-HPLC with a photodiode array detector was used for the determination of flavonoids in fruits of Peucedanum alsaticum. Three compounds were identified: a kaempherol derivative (astragalin), and two quercetin
Choong Je Ma et al.
Journal of enzyme inhibition and medicinal chemistry, 24(3), 676-679 (2008-10-01)
The n-butanol (n-BuOH) fraction of Orostachys japonicus A. Berger (Crassulaceae) significantly inhibited calpain activity. Through the activity-guided isolation from the n-BuOH fraction, herbacetin 8-O-alpha-D-ribopyranoside (1), kaempferol (2), quercetin (3), afzelin (4), astragalin (5), isoquercetin (6) and quercitrin (7) were obtained.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service