Skip to Content
Merck
All Photos(1)

Key Documents

42965

Sigma-Aldrich

Tramadol hydrochloride

≥99.0% (HPLC)

Synonym(s):

Tramadol HCl, (±)-cis-2-(Dimethylaminomethyl)-1-(3-methoxyphenyl)cyclohexanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H25NO2 · HCl
CAS Number:
Molecular Weight:
299.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.25

Assay

≥99.0% (HPLC)

form

powder

drug control

USDEA Schedule IV

mp

178-181 °C

SMILES string

Cl.COc1cccc(c1)[C@@]2(O)CCCC[C@@H]2CN(C)C

InChI

1S/C16H25NO2.ClH/c1-17(2)12-14-7-4-5-10-16(14,18)13-8-6-9-15(11-13)19-3;/h6,8-9,11,14,18H,4-5,7,10,12H2,1-3H3;1H/t14-,16+;/m1./s1

InChI key

PPKXEPBICJTCRU-XMZRARIVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tramadol hydrochloride has been used to study the tramadol biosynthesis route in plants.[1]

Biochem/physiol Actions

Tramadol hydrochloride (HCl) inhibits the reuptake of norepinephrine and serotonin in μ-opioid receptors in rat models in vitro.[2][3]
Opioid analgesic; P450 substrate.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Driessen et al.
British journal of pharmacology, 108(3), 806-811 (1993-03-01)
1. The centrally acting analgesic, tramadol, has low affinity for opioid receptors and therefore presumably other mechanisms of analgesic action. Neurotransmitter release and uptake experiments were used to characterize the effects of tramadol on the central noradrenergic and dopaminergic systems.
Howard S Smith
Mayo Clinic proceedings, 84(7), 613-624 (2009-07-02)
Clinicians understand that individual patients differ in their response to specific opioid analgesics and that patients may require trials of several opioids before finding an agent that provides effective analgesia with acceptable tolerability. Reasons for this variability include factors that
Arshi Kizilbash et al.
Journal of pain research, 7, 149-161 (2014-04-09)
Patients with chronic non-malignant pain report impairments of physical, social, and psychological well-being. The goal of pain management should include reducing pain and improving quality of life. Patients with chronic pain require medications that are able to provide adequate pain
Emrah Yuruk et al.
Journal of endourology, 29(4), 463-467 (2014-10-01)
To compare the outcomes of these minimally invasive procedures in this patient population. The database of our institution has been retrospectively reviewed, and medical records of urolithiasis patients with a solitary kidney who underwent flexible ureteroscopy (F-URS) or extracorporeal shock
Esra Eyiiş et al.
Turkish journal of pharmaceutical sciences, 17(6), 586-592 (2021-01-05)
Safe and effective drugs are still lacking for many pain therapies. In recent years, growing interest has been devoted thus on herbal drugs as an option to identify new pain killers. Based on this, extensive researches are carried out on

Questions

  1. we have sealed vial of tramadol hydrochloride 42695 lot number BBCBV6194. its quality relase date is 02 AUG 2017. the vial is sealed and kept in room temperature. how long this would be valid for use

    1 answer
    1. This product is not assigned an expiration date or recommended retest date. Products with no expiration date or recommended retest date should be routinely inspected by customers to ensure they perform as expected. These products are also subject to a one year warranty from the date of shipment. For more information you may access the "Product Dating Information" document under "ADDITIONAL USEFUL DOCUMENTS ABOUT OUR PRODUCTS" at the bottom of the Quality Services page with this link: https://www.sigmaaldrich.com/life-science/quality-and-regulatory-management/quality-services.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service