Skip to Content
Merck
All Photos(1)

Documents

427M-9

Sigma-Aldrich

p27Kip1 (SX53G8) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

SX53G8, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (427M-94)
vial of 0.5 mL concentrate (427M-95)
bottle of 1.0 mL predilute (427M-97)
vial of 1.0 mL concentrate (427M-96)
bottle of 7.0 mL predilute (427M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

Gene Information

General description

p27 is a cyclin-dependent kinase inhibitor which regulates progression from G1 to S phase by binding and inhibiting cyclin-dependent kinases. Low p27 expression has been associated with unfavorable prognosis in renal cell carcinoma, colon carcinoma, small breast carcinomas, non-small-cell lung carcinoma, hepatocellular carcinoma, multiple myeloma, lymph node metastases in Papillary carcinoma of the Thyroid, and more aggressive phenotype in carcinoma of the cervix.

Quality


IVD

IVD

IVD

RUO

Linkage

p27Kip1 Positive Control Slides, Product No. 427S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Esposito et al.
Cancer research, 57(16), 3381-3385 (1997-08-15)
Despite its potential role as a tumor suppressor, p27 gene, a member of the Cip/Kip family of cyclin-dependent kinase inhibitor genes, has never been found mutated in human tumors. We investigated p27 protein expression in a series of 108 non-small
P Tan et al.
Cancer research, 57(7), 1259-1263 (1997-04-01)
Breast carcinomas < or = 1 cm in size (T1a,b) are being detected more frequently as a result of screening. Because traditional prognostic parameters are either lacking (tumor size) or rare (nodal metastases), a marker(s) is needed to identify the
Carolina Armengol et al.
Journal of hepatology, 38(5), 591-597 (2003-04-26)
Alterations in p27(Kip1) (p27) and cyclin E (cycE) expression are found in tumors and are related to poor prognosis. This study assesses the role of these cell cycle regulators in the development of recurrence after surgical resection in 46 cirrhotic
R V Lloyd et al.
The American journal of pathology, 154(2), 313-323 (1999-02-23)
p27kip1 (p27) is a member of the universal cyclin-dependent kinase inhibitor (CDKI) family. p27 expression is regulated by cell contact inhibition and by specific growth factors, such as transforming growth factor (TGF)-beta. Since the cloning of the p27 gene in
A Haitel et al.
Urology, 58(3), 477-481 (2001-09-11)
To determine whether p21, p27, and pRb can predict disease progression in clear cell renal cell carcinoma. The expression of three negative regulators of the cell cycle, the retinoblastoma gene product (pRb), the WAF1/Cip1 gene product (p21), and the Kip1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service