Skip to Content
Merck
All Photos(1)

Key Documents

42605

Sigma-Aldrich

FragIT MidiSpin Kit

for cleaving and purifying up to 10mg IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

storage temp.

2-8°C

General description

1 ml settled resin of FabRICATOR (IdeS) immobilized on highly cross-linked agarose and 1 ml CapureSelect IgG-Fc affinity ligand immobilized on agarose. Agarose resins are suspended in 20% ethanol/water (V/V)

Application

FragIT MidiSpin Kit (cleaving and purifying up to 10mg IgG) can be used for antibody characterization.

Biochem/physiol Actions

IgG fragmentation is accompanying with adverse consequences like low yield, time consuming optimization and operation at low pH with loss of immunoreactivity. These consequences can be overcome with FragIT MidiSpin Kit, which comprises of pre-filled Fabricator enzyme covalently coupled to agarose beads that facilitates the cleavage of IgG (cleaving and purifying up to 10mg IgG) to generate pure F(ab′)2 and Fc fragments. FabRICATOR is a digestive enzyme that digests IgG specifically below the hinge region so there is no risk of over digestion if incubation time exceeds.[1]

Unit Definition

1 ml of settled resin fragments ≥ 95% of 5 mg of human IgG in 15 minutes at 25°C, pH 6.6 as monitored by SDS-PAGE.

Analysis Note

Enzymatic activity: corresponds to requirements

Legal Information

FragIT is a trademark of Genovis AB

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lukmanee Tradtrantip et al.
Molecular pharmacology, 83(6), 1268-1275 (2013-04-11)
Neuromyelitis optica (NMO) is an inflammatory demyelinating disease of the central nervous system caused by binding of pathogenic IgG autoantibodies (NMO-IgG) to astrocyte water channel aquaporin-4 (AQP4). Astrocyte damage and downstream inflammation require NMO-IgG effector function to initiate complement-dependent cytotoxicity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service