Skip to Content
Merck
All Photos(1)

Key Documents

42129

Lathosterolamide MGI-39

≥97.0% (HPLC)

Synonym(s):

(3S,20S)-20-N-(2-Methylpropyl)carbamoylpregn-7-en-3β-ol, (3S,20S)-3β-Hydroxy-N-(2-methylpropyl)-7-pregnene-20-carboxamide, Lathosterol Oxidase Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H43NO2
CAS Number:
Molecular Weight:
401.63
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

biological source

synthetic

Assay

≥97.0% (HPLC)

form

crystals and lumps

storage temp.

2-8°C

SMILES string

CC(C)CNC(=O)[C@@H](C)[C@H]1CC[C@H]2C3=CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

InChI

1S/C26H43NO2/c1-16(2)15-27-24(29)17(3)21-8-9-22-20-7-6-18-14-19(28)10-12-25(18,4)23(20)11-13-26(21,22)5/h7,16-19,21-23,28H,6,8-15H2,1-5H3,(H,27,29)/t17-,18-,19-,21+,22-,23-,25-,26+/m0/s1

InChI key

HUMXZYXRXDDZOG-LFZVSNMSSA-N

Biochem/physiol Actions

Lathosterolamide MGI-39 is an inhibitor of lathosterol oxidase which converts 5-α-cholest-7-en-3-β-ol to cholesta-5,7-dien-3β-ol.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Giera et al.
Steroids, 73(3), 299-308 (2008-01-01)
Inhibition of cholesterol biosynthesis offers the opportunity for treatment of cardiovascular diseases. Numerous enzymes are involved in the post-squalene part of this biosynthesis, and selective inhibitors for almost all of the enzymes involved there have been described in literature. The
Carolina C Sánchez-Martín et al.
Cancer research, 67(7), 3379-3386 (2007-04-06)
Cholesterol metabolism is particularly active in malignant, proliferative cells, whereas cholesterol starvation has been shown to inhibit cell proliferation. Inhibition of enzymes involved in cholesterol biosynthesis at steps before the formation of 7-dehydrocholesterol has been shown to selectively affect cell
Ralf Klingenstein et al.
Journal of neurochemistry, 98(3), 748-759 (2006-06-06)
Prion diseases are invariably fatal, neurodegenerative diseases transmitted by an infectious agent, PrPSc, a pathogenic, conformational isoform of the normal prion protein (PrPC). Heterocyclic compounds such as acridine derivatives like quinacrine abolish prion infectivity in a cell culture model of
Ken'ichi Hagiwara et al.
Biological & pharmaceutical bulletin, 30(4), 835-838 (2007-04-06)
In prion diseases, the normal cellular form of prion protein (PrP(C)) is converted into the disease-associated isoforms (PrP(Sc)) which accumulate in the infected tissues. Although the precise mechanism of this conversion remains unsolved, drugs of various categories have been reported
Carlos Fernández et al.
Journal of lipid research, 46(5), 920-929 (2005-02-03)
Cholesterol is a major lipid component of the plasma membrane in animal cells. In addition to its structural requirement, cholesterol is essential in cell proliferation and other cell processes. The aim of the present study was to elucidate the stringency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service