Skip to Content
Merck
All Photos(1)

Documents

38361

Sigma-Aldrich

Abberior® STAR 440SXP, maleimide

for long Stokes STED and 2-color STED application

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352111
NACRES:
NA.32

Assay

≥80.0% (degree of coupling)

mol wt

Mw 622.6 g/mol

solubility

DMF: 1 mg/mL, clear

fluorescence

λex 437 nm; λem 515 nm±5 nm in PBS, pH 7.4

storage temp.

−20°C

General description

Absorption Maximum, λmax: 433 nm (MeOH)
432 nm (PBS, pH 7.4)
Extinction Coefficient, ε(λmax): 36,000 M-1cm-1 (MeOH)
33,000 M-1cm-1 (PBS, pH 7.4)
Correction Factor, CF260 = ε260/εmax: 0.47 (PBS, pH 7.4)
Correction Factor, CF280 = ε280/εmax: 0.31 (PBS, pH 7.4)
Fluorescence Maximum, λfl: 502 nm (MeOH),
511 nm (PBS, pH 7.4)
Recommended STED Wavelength, λSTED: 590 − 620 nm
Fluorescence Quantum Yield, η: 0.57 (PBS, pH 7.4)
Fluorescence Lifetime, τ: 3.7 ns (PBS, pH 7.4)

Application

Abberior® STAR 440SXP has been conjugated with secondary anti-mouse antibody for dual colour STED (stimulated emission depletion) microscopy. Again, it has been labelled with secondary antibody for STED microscopy in primary hippocampal cells prepared from E18 Sprague Dawley embryos.

Suitability

Designed and tested for fluorescent super-resolution microscopy

Legal Information

abberior is a registered trademark of Abberior GmbH

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Westin et al.
BMC neuroscience, 15, 45-45 (2014-03-29)
Norbin is a neuron-specific, cytosolic protein that interacts with the metabotropic glutamate receptor 5 (mGluR5) and has a profound impact on mGluR5 signaling. Yet, little is known about its synaptic distribution. Here we have analyzed the spatial relationship between Norbin
I-Hsuan Wang et al.
Cell host & microbe, 14(4), 468-480 (2013-10-22)
Viral DNA trafficking in cells has large impacts on physiology and disease development. Current methods lack the resolution and accuracy to visualize and quantify viral DNA trafficking at single-molecule resolution. We developed a noninvasive protocol for accurate quantification of viral DNA-genome
Tim Grotjohann et al.
Nature, 478(7368), 204-208 (2011-09-13)
Lens-based optical microscopy failed to discern fluorescent features closer than 200 nm for decades, but the recent breaking of the diffraction resolution barrier by sequentially switching the fluorescence capability of adjacent features on and off is making nanoscale imaging routine. Reported
T A Klar et al.
Optics letters, 24(14), 954-956 (2007-12-13)
We overcame the resolution limit of scanning far-field fluorescence microscopy by disabling the fluorescence from the outer part of the focal spot. Whereas a near-UV pulse generates a diffraction-limited distribution of excited molecules, a spatially offset pulse quenches the excited
S W Hell et al.
Optics letters, 19(11), 780-782 (1994-06-01)
We propose a new type of scanning fluorescence microscope capable of resolving 35 nm in the far field. We overcome the diffraction resolution limit by employing stimulated emission to inhibit the fluorescence process in the outer regions of the excitation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service