Skip to Content
Merck
All Photos(2)

Key Documents

36783

Supelco

Fluazifop-butyl

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20F3NO4
CAS Number:
Molecular Weight:
383.36
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

CCCCOC(=O)C(C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1

InChI

1S/C19H20F3NO4/c1-3-4-11-25-18(24)13(2)26-15-6-8-16(9-7-15)27-17-10-5-14(12-23-17)19(20,21)22/h5-10,12-13H,3-4,11H2,1-2H3

InChI key

VAIZTNZGPYBOGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N W McCracken et al.
Biochemical pharmacology, 45(1), 31-36 (1993-01-07)
In the present study, the distribution and nature of esterases in the rat which hydrolysed fluazifop-butyl, carbaryl, paraoxon and phenylacetate were investigated. Vmax and Km values for the hydrolysis reactions were determined. Fluazifop-butyl was hydrolysed to fluazifop by rat liver
Robin J Blake et al.
Pest management science, 68(3), 412-421 (2011-10-06)
The selective graminicide fluazifop-P-butyl is used for the control of grass weeds in dicotyledonous crops, and commonly applied in amenity areas to reduce grass productivity and promote wildflower establishment. However, evidence suggests that fluazifop-P-butyl might also have phytotoxic effects on
J D Ramsey et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 23(2), 230-236 (1994-08-01)
The predictive accuracy of in vitro measurements in estimating dermal absorption has been evaluated in rat and human skin using fluazifop-butyl (FB), a lipophilic model compound, at dosage rates of 2.5, 25, and 250 micrograms/cm2. In vitro studies used rat
High-performance liquid chromatographic determination of Fluazifop-butyl and Fluazifop in soil and water.
M Negre et al.
Journal of chromatography, 387, 541-545 (1987-01-30)
J M Rawlings et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 23(1), 93-100 (1994-07-01)
One important factor which may influence the extent and rate of percutaneous absorption is the dosing vehicle. The purpose of the experiments described was to compare the effect of dosing vehicles of different polarities on the absorption of two herbicides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service