Skip to Content
Merck
All Photos(1)

Key Documents

36098

Sigma-Aldrich

Anti-Rabbit IgG F(ab′)2 fragment - Atto 488, whole molecule antibody produced in goat

1 mg/mL protein

Synonym(s):

Atto 488-[F(ab′)2 fragment goat-Anti-rabbit IgG], whole molecule

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$373.00

$373.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 ML
$373.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$373.00


Estimated to ship on29 May 2025


conjugate

Atto 488 conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

contains

50% glycerol as stabilizer

species reactivity

rabbit

concentration

1 mg/mL protein

fluorescence

λex 500 nm; λem 522 nm in PBS

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Goat anti-rabbit IgGs associate with rabbit IgGs.

Immunogen

rabbit IgG F(ab′)2 fragment

Application

Atto 488-goat anti-rabbit IgG F(ab′)2 fragment, whole molecule antibody can be used for fluorescence-based immunoassays.

Physical form

Atto 488 F(ab)′2 fragment goat anti-rabbit IgG (whole molecule) is provided in unit sizes of 1 ml as 1mg/mL solutions in 0.1 M sodium phosphate, 0.1 M NaCl, pH 7.5, containing 5 mM sodium azide as a preservative.

Preparation Note

F(ab)′2 fragment goat anti-rabbit is conjugated to Atto 488 NHS and purified by gel permeation chromatography and dialysis to remove unbound dye.

Analysis Note

Unconjugated dye ≤5% of total fluorescence; dye-to protein ratio ≥2

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janosch P Heller et al.
Methods in molecular biology (Clifton, N.J.), 1938, 69-84 (2019-01-09)
In recent years it has become apparent that astroglia are not only essential players in brain development, homeostasis, and metabolic support but are also important for the formation and regulation of synaptic circuits. Fine astrocytic processes that can be found in
Shiqi Ma et al.
Neoplasia (New York, N.Y.), 21(4), 343-352 (2019-03-05)
There are seven ligands for the epidermal growth factor receptor (EGFR) ErbB1 and two ligands for ErbB3. EGFR can form a homodimer or a heterodimer with ErbB3. In this study, we investigated whether homodimers or heterodimers, and which ligand, play

Articles

Immunoblotting (Western blot transfer) is a common technique in modern proteomics research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service