Skip to Content
Merck
All Photos(2)

Documents

3514-05A

Sigma-Aldrich

Human Carotid Artery Smooth Muscle Cells: HCtASMC, adult

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human carotid artery (normal)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

23

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

stroke; cardiovascular diseases

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Human Carotid Artery Smooth Muscle Cells (HCtASMC) provide a useful in vitro system to study all aspects of cardiovascular function and disease, especially those related to mechanisms of hyperplasia and hypertrophy of intimal smooth muscle cells leading to vascular occlusion in atherosclerosis and stent restenosis.Characterization: positive for smooth muscle cell specific alpha-actin expression.

Cell Line Origin

Artery

Application

cardiovascluar function, transfer of oxygenated blood to skull, brain, neck, face, effects of drugs

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 10 doublings

Subculture Routine

Please refer to the HCtASMC Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Till Seime et al.
Cells, 10(6) (2021-06-03)
Calcification is a prominent feature of late-stage atherosclerosis, but the mechanisms driving this process are unclear. Using a biobank of carotid endarterectomies, we recently showed that Proteoglycan 4 (PRG4) is a key molecular signature of calcified plaques, expressed in smooth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service