338R-2
TdT (EP266) Rabbit Monoclonal Primary Antibody
Synonym(s):
Terminal Deoxynucleotidyl Transferase
Select a Size
Select a Size
About This Item
Recommended Products
biological source
rabbit
Quality Level
100
500
conjugate
unconjugated
antibody form
culture supernatant
antibody product type
primary antibodies
clone
EP266, monoclonal
description
For In Vitro Diagnostic Use in Select Regions
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (338R-24)
vial of 0.1 mL concentrate Research Use Only (338R-24-RUO)
vial of 0.5 mL concentrate (338R-25)
vial of 1.0 mL concentrate (338R-26)
vial of 1.0 mL concentrate Research Use Only (338R-26-RUO)
vial of 1.0 mL pre-dilute Research Use Only (338R-27-RUO)
vial of 1.0 mL pre-dilute ready-to-use (338R-27)
vial of 7.0 mL pre-dilute ready-to-use (338R-28)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (338R-28-RUO)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:200 (concentrated)
isotype
IgG
control
thymus
shipped in
wet ice
storage temp.
2-8°C
visualization
nuclear
Gene Information
human ... DNTT(1791)
General description
Quality
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Linkage
Physical form
Preparation Note
Note: This requires a keycode which can be found on your packaging or product label.
Download the latest released IFU
Note: This IFU may not apply to your specific product lot.
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service