Skip to Content
Merck
All Photos(1)

Key Documents

29447

Sigma-Aldrich

(24S)-24,25-Dihydroxyvitamin D3

≥95.0% (HPLC)

Synonym(s):

(24S)-24,25-Dihydroxycholecalciferol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H44O3
CAS Number:
Molecular Weight:
416.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥95.0% (HPLC)

form

powder or crystals

color

colorless to white

mp

114.6 °C

storage temp.

−20°C

SMILES string

C[C@H](CC[C@H](O)C(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C27H44O3/c1-18-8-12-22(28)17-21(18)11-10-20-7-6-16-27(5)23(13-14-24(20)27)19(2)9-15-25(29)26(3,4)30/h10-11,19,22-25,28-30H,1,6-9,12-17H2,2-5H3/b20-10+,21-11-/t19-,22+,23-,24+,25+,27-/m1/s1

InChI key

FCKJYANJHNLEEP-WQUHCOROSA-N

Biochem/physiol Actions

Cholecalciferol is an inactive form of vitamin D3 which undergoes various levels of hydroxylation to form active vitamin D3 analogs. 1α-Hydroxyvitamin D3 (alfacalcidol) is a synthetic analog that is metabolized to 1,25-dihydroxycholecalciferol, the biologically active form of vitamin D3. Other analogues of cholecalciferol result from different hydroxylations. 24S,25-Dihydroxyvitamin D3 should not be confused with 24R,25-Dihydroxyvitamin D3.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H L Henry et al.
The Journal of nutrition, 106(6), 724-734 (1976-06-01)
The ability of 24R, 25- and 24S, 25-dihydroxycholecalciferol to stimulate intestinal calcium transport and bone calcium mobilization in chicks was measured. Enhancement of intestinal calcium transport by 325 or 130 nmoles of either compound was maximal by 24 hours. The
José Pérez Sestelo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 8(12), 2747-2752 (2002-10-24)
Vitamin D3 active metabolites 24R,25-(OH)2-D3, 24S,25-(OH)2-D3, and 1 alpha, 24R,25-(OH)3-D3 were synthesized by a convergent and stereoselective approach. In the synthetic route, the stereogenic center at C-24 was generated through ultrasonically induced aqueous conjugate addition of iodide 6 to Seebach's
S Ishizuka et al.
Archives of biochemistry and biophysics, 234(1), 97-104 (1984-10-01)
Tritium-labeled 24,25-dihydroxyvitamin D3 was prepared both in vitro, by using chick kidney homogenates, and in vivo in rats from [26,27-methyl-3H]25-hydroxyvitamin D3. These compounds were mixed with synthetic 24(R),25- and 24(S),25-dihydroxyvitamin D3, converted to the corresponding trimethylsilyl ether derivatives, and analyzed
Anjali Verma et al.
Steroids, 150, 108447-108447 (2019-07-16)
Vitamin D has long been prescribed as a supplement to breast cancer patients. This is partially motivated by data indicating that low serum vitamin D, measured as 25-hydroxyvitamin D3 [25(OH)D3], is associated with worsened cancer prognosis and decreased survival rates
Anjali Verma et al.
Molecular cancer research : MCR, 19(1), 99-111 (2020-10-22)
Vitamin D3 and its metabolites have antitumorigenic properties in vitro and in vivo; however, clinical trials and retrospective studies on the effectiveness of vitamin D3 oral supplementation against cancer have been inconclusive. One reason for this may be that clinical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service