Skip to Content
Merck
All Photos(1)

Documents

255M-1

Sigma-Aldrich

Galectin-3 (9C4) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

9C4, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (255M-14)
vial of 0.5 mL concentrate (255M-15)
bottle of 1.0 mL predilute (255M-17)
vial of 1.0 mL concentrate (255M-16)
bottle of 7.0 mL predilute (255M-18)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100

isotype

IgG1

control

papillary carcinoma of thyroid

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic, nuclear

Gene Information

human ... LGALS3(3958)

General description

Galectin-3 is a 31 kD beta-galactosidase binding lectin. It has been associated with binding to the basement membrane glycoprotein laminin. Anti-Galectin-3 has been demonstrated to be valuable in differentiating between benign and malignant thyroid neoplasms in both histologic sections and fine needle aspiration biopsy material. Anti-Galectin-3 antibody has also been useful in identifying anaplastic large cell lymphoma.

Quality


IVD

IVD

IVD

RUO

Linkage

Galectin-3 Positive Control Slides, Product No. 255S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K N Konstantinov et al.
The American journal of pathology, 148(1), 25-30 (1996-01-01)
Galectin-3 is a member of a newly named family of beta-galactoside-binding animal lectins, which has been described with a number of possible important biological functions, including the regulation of cell growth and association with tumor transformation. This protein has a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service