Skip to Content
Merck
All Photos(1)

Key Documents

24660C

SAFC

EX-CELL® Sp2/0 Serum-Free Medium for Sp2/0 Cells

Animal-component free, Protein free, Chemically defined, without L-glutamine, without sodium bicarbonate, dry powder, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$147.00
5 L
$428.00
10 L
$733.00
50 L
$2,646.00

$147.00


Check Cart for Availability


Select a Size

Change View
1 L
$147.00
5 L
$428.00
10 L
$733.00
50 L
$2,646.00

About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.71

$147.00


Check Cart for Availability

Quality Level

description

for research or for further manufacturing use

form

dry powder

technique(s)

cell culture | mammalian: suitable

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Quantity

Formulated to contain 17.3 grams of powder per liter of medium.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Eukaryotic cell lines aid in complex post-translational modifications and folding of recombinant proteins. Find suitable recombinant protein production cell lines.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service